Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPLP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103540 Supplier Invitrogen™ Supplier No. PA5103540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RPLP1.

Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein LP1 (ribosomal protein, large, P1), also known as 60S acidic ribosomal protein P1, RPP1 or RPLP1, is a 114 amino acid acidic protein that belongs to the ribosomal protein L12P family. A key component during the elongation step of protein synthesis, Ribosomal Protein LP1 forms a pentameric complex (in conjunction with LP2 and LP0) that regulates ribosome assembly and plays a role in translation initiation. Like other mammalian ribosomal proteins, Ribosomal Protein LP1 exists as multiple processed pseudogenes that are found throughout the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPLP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPLP1
Gene Accession No. P05386, P19944, P47955
Gene Alias 2410042H16Rik; 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; Arpp1; C430017H15Rik; L RPP1; Large ribosomal subunit protein P1; LP1; P1; ribosomal protein lateral stalk subunit P1; ribosomal protein, large, P1; RPLP1; RPP1; RRP1
Gene Symbols RPLP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPLP1(Accession P05386), corresponding to amino acid residues G36-A86.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140661, 56040, 6176
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.