Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPLP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121030
This item is not returnable.
View return policy
Description
Positive test controls include: SW480, Mouse spleen, Mouse lung, Mouse brain, Rat spleen, Rat lung, Rat brain. Immunogen sequence: MASVSELACI YSALILHDDE VTVTEDKINA LIKAAGVNVE PFWPGLFAKA LANVNIGSLI CNVGAGGPAP AAGAAPAGGP APSTAAAPAE EKKVEAKKEE SEESDDDMGF GLFD.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein LP1 (ribosomal protein, large, P1), also known as 60S acidic ribosomal protein P1, RPP1 or RPLP1, is a 114 amino acid acidic protein that belongs to the ribosomal protein L12P family. A key component during the elongation step of protein synthesis, Ribosomal Protein LP1 forms a pentameric complex (in conjunction with LP2 and LP0) that regulates ribosome assembly and plays a role in translation initiation. Like other mammalian ribosomal proteins, Ribosomal Protein LP1 exists as multiple processed pseudogenes that are found throughout the genome.
Specifications
| RPLP1 | |
| Polyclonal | |
| Unconjugated | |
| RPLP1 | |
| 2410042H16Rik; 60S acidic ribosomal protein P1; acidic ribosomal phosphoprotein P1; Arpp1; C430017H15Rik; L RPP1; Large ribosomal subunit protein P1; LP1; P1; ribosomal protein lateral stalk subunit P1; ribosomal protein, large, P1; RPLP1; RPP1; RRP1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140661, 336719, 56040, 6176 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05386, P19944, P47955 | |
| RPLP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-114 of human RPLP1 (NP_000994.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction