Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPLP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562820 Supplier Invitrogen™ Supplier No. PA562820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRYVASYLLA ALGGNSSPSA KDIKKILDSV GIEADDDRLN KVIS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 95%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit.Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal phosphoprotein that is a component of the 60S subunit. The protein, which is a functional equivalent of the E. coli L7/L12 ribosomal protein, belongs to the L12P family of ribosomal proteins.It plays an important role in the elongation step of protein synthesis. Unlike most ribosomal proteins, which are basic, the encoded protein is acidic. Its C-terminal end is nearly identical to the C-terminal ends of the ribosomal phosphoproteins P0 and P1. The P2 protein can interact with P0 and P1 to form a pentameric complex consisting of P1 and P2 dimers, and a P0 monomer. The protein is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. (provided by RefSeq, Jul 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPLP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPLP2
Gene Accession No. P05387
Gene Alias 2700049I22Rik; 60S acidic ribosomal protein P2; acidic ribosomal phosphoprotein P2; D11S2243E; L RP D11S2243E; Large ribosomal subunit protein P2; LP2; P2; Renal carcinoma antigen NY-REN-44; ribosomal protein lateral stalk subunit P2; ribosomal protein P2; ribosomal protein, large P2; ribosomal protein, large, P2; RP2; Rplp2; RPP2
Gene Symbols RPLP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPLP2. Recombinant protein control fragment (Product #RP-101680).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.