Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
rpmB/L28 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323513JF646
Description
rpmB/L28 Polyclonal antibody specifically detects rpmB/L28 in E. coli samples. It is validated for Western Blot, ELISASpecifications
rpmB/L28 | |
Polyclonal | |
Western Blot, ELISA | |
50S ribosomal subunit protein L28, ECK3627, Large ribosomal subunit protein bL28, rpmB | |
Antibody was raised against an 19 amino acid peptide near the carboxy terminus of E. coli rpmB/L28. The immunogen is located within the last 50 amino acids of rpmB/L28. | |
0.1 mL | |
Primary | |
E. coli | |
Purified |
Western Blot, ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
946941 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction