Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
rpmB/L28 Antibody - N-Terminus - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$170.70 - $413.00
Specifications
Antigen | rpmB/L28 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5 ug/ml, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
rpmB/L28 Polyclonal antibody specifically detects rpmB/L28 in E. coli samples. It is validated for Western Blot, ELISASpecifications
rpmB/L28 | |
Western Blot 0.5 ug/ml, ELISA | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
946941 | |
IgG | |
Affinity purified |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
E. coli | |
50S ribosomal subunit protein L28, ECK3627, Large ribosomal subunit protein bL28, rpmB | |
Antibody was raised against an 15 amino acid peptide near the amino terminus of E. coli rpmB/L28. The immunogen is located within the first 50 amino acids of rpmB/L28. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title