Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
rpmH/L34 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323515B
Description
rpmH/L34 Polyclonal antibody specifically detects rpmH/L34 in E. coli samples. It is validated for Western Blot, ELISASpecifications
rpmH/L34 | |
Polyclonal | |
Western Blot, ELISA | |
50S ribosomal subunit protein L34, ECK3695, Large ribosomal subunit protein bL34, rimA, rpmH, ssaF | |
Antibody was raised against an 17 amino acid peptide near the amino terminus of E. coli rpmH/L34. The immunogen is located within the first 30 amino acids of rpmH/L34. | |
0.1 mL | |
Primary | |
E. coli | |
Purified |
Western Blot, ELISA | |
Biotin | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
948216 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction