Learn More
RPN1 Mouse anti-Canine, Human, Clone: OTI2G1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503654

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombit Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.Specifications
RPN1 | |
Monoclonal | |
Unconjugated | |
RPN1 | |
OST1, RBPH1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
476516, 6184 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2G1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
E2RQ08, P04843 | |
RPN1 | |
Full length human recombit protein of human RPN1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.