Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rpn2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1202941
Description
Rpn2 Polyclonal specifically detects Rpn2 in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Rpn2 | |
Polyclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
26S proteasome non-ATPase regulatory subunit 1, 26S proteasome regulatory subunit RPN2, 26S proteasome regulatory subunit S1, 26S proteasome subunit p112, MGC133040, MGC133041, P112, proteasome (prosome, macropain) 26S subunit, non-ATPase, 1, Rpn2, S1 | |
Rabbit | |
106 kDa | |
100 μg | |
Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway | |
5707 | |
Human, Mouse, Rat, Bovine, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1.0 ug/ml, Immunohistochemistry 15 ug/ml, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
Q99460 | |
PSMD1 | |
Synthetic Peptide: A(929) A H G P K I E E E E Q E P E P P E P F E Y I D D(953) | |
Affinity Purified | |
RUO | |
Primary | |
Detects proteasome 19S subunit S1 from human cells. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction