Learn More
RPN2 Mouse anti-Human, Clone: OTI1G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504604

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.Specifications
RPN2 | |
Monoclonal | |
Unconjugated | |
RPN2 | |
RIBIIR, RPN-II, RPNII, SWP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6185 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P04844 | |
RPN2 | |
Full length human recombit protein of human RPN2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.