Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107017 Supplier Invitrogen™ Supplier No. PA5107017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.

RPP21 is a protein subunit of nuclear ribonuclease P, which processes the 5-prime leader sequence of precursor tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP21
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene Rpp21
Gene Accession No. Q9H633
Gene Alias 0610037N01Rik; C6orf135; Cat60; Flj22638; ribonuclease P 21 subunit; ribonuclease P 21kDa subunit; ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21 subunit; ribonuclease P/MRP 21kDa subunit; ribonuclease P/MRP subunit p21; ribonucleoprotein V; RNaseP protein p21; Rpp21
Gene Symbols Rpp21
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: LVPGLTCTQR QRRCRGQRWT VQTCLTCQRS QRFLNDPGHL LWGDRPEAQL.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.