Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113486
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113486 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113486 Supplier Invitrogen™ Supplier No. PA5113486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPP21 Polyclonal Antibody for Western Blot, ELISA

RPP21 is a protein subunit of nuclear ribonuclease P, which processes the 5-prime leader sequence of precursor tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP21
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Rpp21
Gene Accession No. Q9H633
Gene Alias 0610037N01Rik; C6orf135; Cat60; Flj22638; ribonuclease P 21 subunit; ribonuclease P 21kDa subunit; ribonuclease P protein subunit p21; Ribonuclease P/MRP 21 kDa subunit; ribonuclease P/MRP 21 subunit; ribonuclease P/MRP 21kDa subunit; ribonuclease P/MRP subunit p21; ribonucleoprotein V; RNaseP protein p21; Rpp21
Gene Symbols Rpp21
Host Species Rabbit
Immunogen Recombinant Human Ribonuclease P protein subunit p21 protein (2-154AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79897
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.