Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561369 Supplier Invitrogen™ Supplier No. PA561369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASLSVLKNVP GLAILLSKDA LDPRQPGYQP PNPHPGPSSP PAAPASKRSL GEPAAGEGSA KRSQPEP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Component of ribonuclease P, a ribonucleoprotein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:12003489, PubMed:16723659, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP25
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPP25
Gene Accession No. Q9BUL9
Gene Alias AI851155; ribonuclease P 25 subunit; ribonuclease P 25kDa subunit; ribonuclease P protein subunit p25; ribonuclease P/MRP 25 subunit; ribonuclease P/MRP 25kDa subunit; ribonuclease P/MRP subunit p25; RNase P protein subunit p25; Rpp25
Gene Symbols RPP25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPP25. Recombinant protein control fragment (Product #RP-100769).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.