Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597315 Supplier Invitrogen™ Supplier No. PA597315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MENFRKVRSE EAPAGCGAEG GGPGSGPFAD LAPGAVHMRV KEGSKIRNLM AFATASMAQP ATRAIVFSGC GRATTKTVTC AEILKRRLAG LHQVTRLRYR SVREVWQSLP PGPTQGQTPG EPAASLSVLK NVPGLAILLS KDALDPRQPG YQPPNPHPGP SSPPAAPASK RSLGEPAAGE GSAKRSQPEP GVADEDQTA; Positive Samples: A-375, HeLa, HT-29; Cellular Location: Nucleus.

Component of ribonuclease P, a ribonucleoprotein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:12003489, PubMed:16723659, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP25
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPP25
Gene Accession No. Q91WE3, Q9BUL9
Gene Alias AI851155; ribonuclease P 25 subunit; ribonuclease P 25kDa subunit; ribonuclease P protein subunit p25; ribonuclease P/MRP 25 subunit; ribonuclease P/MRP 25kDa subunit; ribonuclease P/MRP subunit p25; RNase P protein subunit p25; Rpp25
Gene Symbols RPP25
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102614, 54913
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.