Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP25L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566275 Supplier Invitrogen™ Supplier No. PA566275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEHYRKAGSVE LPAPSPMPQL PPDTLEMRVR DGSKIRNLLG LALGRLE Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

May be a component of ribonuclease P or MRP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP25L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPP25L
Gene Accession No. Q8N5L8
Gene Alias 2810432D09Rik; alba-like protein C9orf23; alba-like protein C9orf23 homolog; AW987258; bA296L22.5; C9orf23; RGD1306576; Ribonuclease P protein subunit p25-like protein; ribonuclease P/MRP 25 subunit-like; ribonuclease P/MRP 25kDa subunit-like; ribonuclease P/MRP subunit p25 like; RNase P protein subunit-like p25; Rpp25l; Rpp25-like protein; Rppl25
Gene Symbols RPP25L
Host Species Rabbit
Immunogen Recombinant Human RPP25L. Recombinant protein control fragment (Product #RP-106961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138716
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.