Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPP25L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568087
Description
Predicted to react with Mouse samples.
May be a component of ribonuclease P or MRP.
Specifications
RPP25L | |
Polyclonal | |
Unconjugated | |
RPP25L | |
2810432D09Rik; alba-like protein C9orf23; alba-like protein C9orf23 homolog; AW987258; bA296L22.5; C9orf23; RGD1306576; Ribonuclease P protein subunit p25-like protein; ribonuclease P/MRP 25 subunit-like; ribonuclease P/MRP 25kDa subunit-like; ribonuclease P/MRP subunit p25 like; RNase P protein subunit-like p25; Rpp25l; Rpp25-like protein; Rppl25 | |
Rabbit | |
affinity chromatography | |
RUO | |
138716, 69961 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8N5L8, Q99JH1 | |
RPP25L | |
Synthesized peptide derived from human RPP25L. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction