Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP25L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103588 Supplier Invitrogen™ Supplier No. PA5103588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C9orf23.

May be a component of ribonuclease P or MRP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP25L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPP25L
Gene Accession No. Q8N5L8, Q99JH1
Gene Alias 2810432D09Rik; alba-like protein C9orf23; alba-like protein C9orf23 homolog; AW987258; bA296L22.5; C9orf23; RGD1306576; Ribonuclease P protein subunit p25-like protein; ribonuclease P/MRP 25 subunit-like; ribonuclease P/MRP 25kDa subunit-like; ribonuclease P/MRP subunit p25 like; RNase P protein subunit-like p25; Rpp25l; Rpp25-like protein; Rppl25
Gene Symbols RPP25L
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPP25L(Accession Q8N5L8), corresponding to amino acid residues C131-S163.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138716, 69961
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.