Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ RPP38 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111212
Description
Immunogen sequence: FVDEVRAIIP RVPSLSVPWL QDRIEDSGEN LETEPLESQD RELLDTSFED LSKPKRKLAD GRQASVTLQP LKIKKLIPNP NKIRKPPKS.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.
            Specifications
| RPP38 | |
| Polyclonal | |
| Unconjugated | |
| RPP38 | |
| C330006A15Rik; ribonuclease P protein subunit p38; ribonuclease P/MRP 38 subunit; ribonuclease P/MRP 38kDa subunit; ribonuclease P/MRP subunit p38; RNaseP protein p38; RPP38 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10557 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78345 | |
| RPP38 | |
| Recombinant Protein corresonding to Human RPP38. Recombinant protein control fragment (Product #RP-100663). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction