Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPP40 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23826025UL
Description
RPP40 Polyclonal specifically detects RPP40 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RPP40 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
O75818 | |
RPP40 | |
This antibody was developed against a recombinant protein corresponding to amino acids: YNYRVSFLIPECGILSEELKNLVMNTGPYYFVKNLPLHELITPEFISTFIKKGSCYALTYNTHIDEDNTVALLPNGKL | |
25 μL | |
Core ESC Like Genes, Stem Cell Markers | |
10799 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
bA428J1.3,40kD subunit, EC 3.1.26.5, ribonuclease P/MRP 40kDa subunit, RNaseP protein p40 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction