Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP40 Polyclonal Antibody

Catalog No. PIPA535968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535968 Supplier Invitrogen™ Supplier No. PA535968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (82%), Rat (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP40
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene RPP40
Gene Accession No. O75818
Gene Alias bA428J1.3; D8Bwg1265e; ribonuclease P (40 kD); ribonuclease P 40 subunit; ribonuclease P 40kDa subunit; ribonuclease P protein subunit p40; ribonuclease P, 40kD subunit; ribonuclease P/MRP 40 subunit; ribonuclease P/MRP 40kDa subunit; ribonuclease P/MRP subunit p40; ribonuclease P1; RNase P subunit 1; RNaseP protein p40; Rnasep1; RPP40
Gene Symbols RPP40
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RPP40. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.