Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPP40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557364
Description
Immunogen sequence: YNYRVSFLIP ECGILSEELK NLVMNTGPYY FVKNLPLHEL ITPEFISTFI KKGSCYALTY NTHIDEDNTV ALLPNGKL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends.
Specifications
| RPP40 | |
| Polyclonal | |
| Unconjugated | |
| RPP40 | |
| bA428J1.3; D8Bwg1265e; ribonuclease P (40 kD); ribonuclease P 40 subunit; ribonuclease P 40kDa subunit; ribonuclease P protein subunit p40; ribonuclease P, 40kD subunit; ribonuclease P/MRP 40 subunit; ribonuclease P/MRP 40kDa subunit; ribonuclease P/MRP subunit p40; ribonuclease P1; RNase P subunit 1; RNaseP protein p40; Rnasep1; RPP40 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75818 | |
| RPP40 | |
| Recombinant protein corresponding to Human RPP40. Recombinant protein control fragment (Product #RP-102675). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction