Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPP40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557365
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557365 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557365 Supplier Invitrogen™ Supplier No. PA557365
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RALELFDWLG AVFSNVDLNN EPNNFISTYC CPEPSTVVAK AYLCTITGFI LPEKICLLLE HLCHYFDEPK LAPWVTLSVQ Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPP40
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPP40
Gene Accession No. O75818
Gene Alias bA428J1.3; D8Bwg1265e; ribonuclease P (40 kD); ribonuclease P 40 subunit; ribonuclease P 40kDa subunit; ribonuclease P protein subunit p40; ribonuclease P, 40kD subunit; ribonuclease P/MRP 40 subunit; ribonuclease P/MRP 40kDa subunit; ribonuclease P/MRP subunit p40; ribonuclease P1; RNase P subunit 1; RNaseP protein p40; Rnasep1; RPP40
Gene Symbols RPP40
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPP40. Recombinant protein control fragment (Product #RP-94809).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.