Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPRD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559219 Supplier Invitrogen™ Supplier No. PA559219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIVEAFKHVS SETDESCKKH LGRVLSIWEE RSVYENDVLE QLKQALYGDK KPRKRTYEQI KVDENENCSS LGSPSEPPQT L Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

P15RS is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15 and may have a role in cell cycle regulation (Liu et al., 2002 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPRD1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPRD1A
Gene Accession No. Q96P16
Gene Alias C77387; CDKN2B-related protein; cyclin-dependent kinase 2B-inhibitor-related protein; cyclin-dependent kinase inhibitor 2B-related protein; Cyclin-dependent kinase inhibitor 2B-related protein (p15INK4B-related protein); HsT3101; Kiaa4077; mKIAA4077; p15INK4B-related protein; P15RS; regulation of nuclear pre-mRNA domain containing 1A; regulation of nuclear pre-mRNA domain-containing protein 1A; RGD1307374; Rprd1a
Gene Symbols RPRD1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPRD1A. Recombinant protein control fragment (Product #RP-98274).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55197
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.