Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPRD1B Antibody (OTI1C7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273945UV
Description
RPRD1B Monoclonal specifically detects RPRD1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RPRD1B | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RPRD1B | |
Full length recombinant protein of human RPRD1B (NP_067038) produced in HEK293T cells. | |
0.1 mL | |
58490.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C7 | |
Western Blot, Immunohistochemistry | |
Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction