Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPRD1B Monoclonal Antibody (08)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529526
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RPRD1B.
Interacts with phosphorylated C-terminal heptapeptide repeat domain (CTD) of the largest RNA polymerase II subunit POLR2A, and participates in dephosphorylation of the CTD by RPAP2. Transcriptional regulator which enhances expression of CCND1. Promotes binding of RNA polymerase II to the CCDN1 promoter and to the termination region before the poly-A site but decreases its binding after the poly-A site. Prevents RNA polymerase II from reading through the 3' end termination site and may allow it to be recruited back to the promoter through promotion of the formation of a chromatin loop. Also enhances the transcription of a number of other cell cycle-related genes including CDK2, CDK4, CDK6 and cyclin-E but not CDKN1A, CDKN1B or cyclin-A. Promotes cell proliferation. [UniProt].
Specifications
RPRD1B | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NQG5 | |
RPRD1B | |
Recombinant Human RPRD1B Protein (Met1-Asp326). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin) | |
08 | |
Unconjugated | |
RPRD1B | |
2610304G08Rik; 2810446G03Rik; C20orf77; Cell cycle-related and expression-elevated protein in tumor; cell-cycle related and expression-elevated protein in tumor; CREPT; dJ1057B20.2; NET60; regulation of nuclear pre-mRNA domain containing 1B; regulation of nuclear pre-mRNA domain-containing protein 1B; RGD1304782; Rprd1b | |
Mouse | |
Protein A | |
RUO | |
58490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction