Learn More
Invitrogen™ RPRD1B Monoclonal Antibody (OTI1C7)

Description
RPRD1B Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RPRD1B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1C7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPRD1B |
| Gene Accession No. | Q9NQG5 |
| Gene Alias | 2610304G08Rik; 2810446G03Rik; C20orf77; Cell cycle-related and expression-elevated protein in tumor; cell-cycle related and expression-elevated protein in tumor; CREPT; dJ1057B20.2; NET60; regulation of nuclear pre-mRNA domain containing 1B; regulation of nuclear pre-mRNA domain-containing protein 1B; RGD1304782; Rprd1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.