Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPRD2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100366AF700
Description
RPRD2 Polyclonal antibody specifically detects RPRD2 in Human samples. It is validated for Western BlotSpecifications
RPRD2 | |
Polyclonal | |
Western Blot | |
HSPC099, KIAA0460FLJ32145, regulation of nuclear pre-mRNA domain containing 2, regulation of nuclear pre-mRNA domain-containing protein 2 | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human KIAA0460 using the numbering given in entry CAH70314.1 (GeneID 23248). | |
0.1 mL | |
DNA Repair | |
23248 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction