Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPRD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570084
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Rat: 86%.
RPRD2 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; RNA polymerase II, holoenzyme; RNA polymerase II complex binding; mRNA 3'-end processing; snRNA transcription by RNA polymerase II.
Specifications
RPRD2 | |
Polyclonal | |
Unconjugated | |
RPRD2 | |
2810036A19Rik; 4930535B03Rik; 6720469I21Rik; AL022841; AL022940; AU021304; BB077382; HSPC099; KIAA0460; LOC100364568; mKIAA0460; regulation of nuclear pre-mRNA domain containing 2; regulation of nuclear pre-mRNA domain containing 2-like; regulation of nuclear pre-mRNA domain-containing protein 2; Rprd2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23248 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VT52 | |
RPRD2 | |
synthetic peptide directed towards the middle region of human RPRD2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction