Learn More
Invitrogen™ RPRD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110324
Description
Immunogen sequence: MAAGGGGGSS KASSSSASSA GALESSLDRK FQSVTNTMES IQGLSSWCIE NKKHHSTIVY HWMKWLRRSA YPHRLNLFYL ANDVIQNCKR KNAIIFRESF ADVLPEAAAL VKDPSVSKSV ERIFKIWEDR NVYPEEMIVA LREALTSTNP KAALKSKIVA EFRSQALIEE LLLYKRSEDQ IELKEKQLST MRVDVCSTET LKCLKDKTGG KKFSKEFEEA SSKLEEFVNG LDKQVKNGPS LTEALENAGI FYEAQYKEVK VVANAYKTFA NRVNNLKKKL DQLKSTLPDP EESPVPSPSM DAPSPTGSES PFQGMGGEES QSPTMESEKS ATPEPVTDNR DVEDMELSDV.
RPRD2 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; RNA polymerase II, holoenzyme; RNA polymerase II complex binding; mRNA 3'-end processing; snRNA transcription by RNA polymerase II.
Specifications
| RPRD2 | |
| Polyclonal | |
| Unconjugated | |
| RPRD2 | |
| 2810036A19Rik; 4930535B03Rik; 6720469I21Rik; AL022841; AL022940; AU021304; BB077382; HSPC099; KIAA0460; LOC100364568; mKIAA0460; regulation of nuclear pre-mRNA domain containing 2; regulation of nuclear pre-mRNA domain containing 2-like; regulation of nuclear pre-mRNA domain-containing protein 2; Rprd2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 75137 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6NXI6 | |
| RPRD2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human RPRD2 (NP_001284603.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.