Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPRD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566413 Supplier Invitrogen™ Supplier No. PA566413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEESPVPSPSM DAPSPTGSES PFQGMGGEES QSPTMESEKS ATPEPVTDNR DVEDMELSDV EDDGSKIIVE DRKEKPAEKS AVSTSVPTKP TENIS Highest antigen sequence identity to the following orthologs - mouse 91%, rat 94%.

RPRD2 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; RNA polymerase II, holoenzyme; RNA polymerase II complex binding; mRNA 3'-end processing; snRNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPRD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPRD2
Gene Accession No. Q5VT52
Gene Alias 2810036A19Rik; 4930535B03Rik; 6720469I21Rik; AL022841; AL022940; AU021304; BB077382; HSPC099; KIAA0460; LOC100364568; mKIAA0460; regulation of nuclear pre-mRNA domain containing 2; regulation of nuclear pre-mRNA domain containing 2-like; regulation of nuclear pre-mRNA domain-containing protein 2; Rprd2
Gene Symbols RPRD2
Host Species Rabbit
Immunogen Recombinant Human RPRD2. Recombinant protein control fragment (Product #RP-107088).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.