Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPRM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587563
Description
Immunogen sequence: MNPALGNQTD VAGLFLANSS EALERAVRCC TQASVVTDDG FAEGGPDERS LYIMRVVQIA VMCVLSLTVV FGIFFLGCNL LIKSEGMINF LVKDRRPSKE VEAVVVGPY; Positive Samples: Mouse pancreas; Cellular Location: Cytoplasm, Membrane, Single-pass membrane protein.
May be involved in the regulation of p53-dependent G2 arrest of the cell cycle. Seems to induce cell cycle arrest by inhibiting CDK1 activity and nuclear translocation of the CDC2 cyclin B1 complex.
Specifications
RPRM | |
Polyclonal | |
Unconjugated | |
RPRM | |
2410012A13Rik; candidate mediator of the p53 dependent G2 arrest; candidate mediator of the p53-dependent G2 arrest; FLJ90327; protein reprimo; Reprimo; reprimo, TP53 dependant G2 arrest mediator candidate; reprimo, TP53 dependent G2 arrest mediator candidate; Rprm | |
Rabbit | |
Affinity chromatography | |
RUO | |
67874 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JJ72 | |
RPRM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-109 of human RPRM (NP_0628191). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction