Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPRM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578538
Description
Positive Control: Target recombinant protein Predicted Reactivity: Mouse (97%), Rat (98%), Rhesus Monkey (100%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in the regulation of p53-dependent G2 arrest of the cell cycle. Seems to induce cell cycle arrest by inhibiting CDK1 activity and nuclear translocation of the CDC2 cyclin B1 complex.
Specifications
RPRM | |
Polyclonal | |
Unconjugated | |
RPRM | |
2410012A13Rik; candidate mediator of the p53 dependent G2 arrest; candidate mediator of the p53-dependent G2 arrest; FLJ90327; protein reprimo; Reprimo; reprimo, TP53 dependant G2 arrest mediator candidate; reprimo, TP53 dependent G2 arrest mediator candidate; Rprm | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NS64 | |
RPRM | |
Full length human RPRM Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction