Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPRML Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30631350UL
Description
RPRML Polyclonal antibody specifically detects RPRML in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
RPRML | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:50-1:200 | |
reprimo-like, Reprimo-Like Protein | |
Produced in rabbits immunized with E. coli-derived Human RPRML fragment. | |
50 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
388394 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction