Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva rProtein A Sepharose™ Fast Flow Affinity Media
GSA_VA

Catalog No. 45002973 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
Binding Capacity:
50 mg Human lgG/mL
50 mg Human lgG/mL medium2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Binding Capacity
45-002-973 200 mL 50 mg Human lgG/mL medium2
45-002-972 25 mL 50 mg Human lgG/mL
45-002-971 5 mL 50 mg Human lgG/mL medium2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-002-973 Supplier Cytiva Supplier No. 17127903
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Designed for purification of monoclonal and polyclonal antibodies at labs and process scales

  • Composed of recombinant protein A coupled with cross-linked 4% agarose base matrix
  • High dynamic binding capacity, single-point attachment of protein A, with high recovery, high purity and low ligand leakage
  • No mammalian components involved during manufacturing process
  • Matrix: 4% cross-linked agarose
  • Average particle size: 90 μm (d50V)
  • Ligand: 6 mg rProtein A/mL medium (E. coli)
  • Binding capacity: approx. 50 mg human IgG/mL media
  • pH stability working range: 3 to 10
  • Pressure/Flow specification: base matrix 150 to 250 cm/hr., 100 kPa, XK 50/60 column, bed

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) For PurifiCatIon of Monoclonal and Polyclonal Antibodies at Labs and Process Scales
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
Storage Requirements 2°C to 8°C
Binding Capacity 50 mg Human lgG/mL medium2
Product Type rProtein A Sepharose Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 60 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 2 to 9
Storage Buffer 20% Ethanol
Ligand Type Recombinant Protein A (E. coli)
Format Bulk Media
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.