Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572303
Description
RPS10 Polyclonal Antibody for Western Blot, ICC/IF, Flow
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein S10 (RPS10) is a 165 amino acid component of the small ribosomal 40S subunit. While generally found in the cytoplasm, RPS10 can localize to the granular component (GC) region of the nucleolus. For this to occur, methylation by PRMT5 is required as well as an interaction with NPS1. Defects in RPS10 are the cause of Diamond-Blackfan anemia type 9 (DBA9). It is a form of Diamond-Blackfan anemia, a congenital non-regenerative hypoplastic anemia that usually presents early in infancy. Diamond-Blackfan anemia is characterized by a moderate to severe macrocytic anemia, erythroblastopenia, and an increased risk of malignancy.
Specifications
RPS10 | |
Polyclonal | |
Unconjugated | |
Rps10 | |
2210402A09Rik; 40S ribosomal protein S10; DBA9; ribosomal protein S10; RP11-375E1__A.4; Rps10; S10; Small ribosomal subunit protein eS10 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
6204, 67097, 81773 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
0.62 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P46783, P63325, P63326 | |
Rps10 | |
KLH conjugated synthetic peptide between 96-129 amino acids from human RPS10. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction