Learn More
Invitrogen™ RPS11 Polyclonal Antibody

Description
Antibody detects endogenous levels of total RPS11.
Specifications
Specifications
| Antigen | RPS11 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RPS11 |
| Gene Accession No. | P62280, P62281, P62282 |
| Gene Alias | 40S ribosomal protein S11; ribosomal protein S11; RPS 11; RPS11; S11; Small ribosomal subunit protein uS17 |
| Gene Symbols | RPS11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.