Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564909
Description
Immunogen sequence: AAGGVMDVNTA LQEVLKTALI HDGLARGIRE AAKALDKRQA HLCVLASNCD EPMYVKLVEA LCAEHQINLI KVDDNKKLGE WVGLCKIDRE GKPRKVVGCS CVVVKDYGKE SQAKDV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S12E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal cancers compared to matched normal colonic mucosa has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS12 | |
Polyclonal | |
Unconjugated | |
RPS12 | |
40S ribosomal protein S12; ribosomal protein S12; Rps12; S12; Small ribosomal subunit protein eS12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6206 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P25398 | |
RPS12 | |
Recombinant Human RPS12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction