Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS14 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222319AF594
Description
RPS14 Polyclonal antibody specifically detects RPS14 in Human, Mouse samples. It is validated for Western BlotSpecifications
RPS14 | |
Polyclonal | |
Western Blot | |
emetine resistance, EMTB40S ribosomal protein S14, ribosomal protein S14 | |
The immunogen this antibody was made to, maps to a region between residue 101 to 151 of human Ribosomal Protein S14 using the numbering given in entry NP_001020242.1 (GeneID 6208). | |
0.1 mL | |
metabolism | |
6208 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction