Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222319
Description
RPS14 Polyclonal specifically detects RPS14 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
RPS14 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
emetine resistance, EMTB40S ribosomal protein S14, ribosomal protein S14 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
NP_001020242.1 | |
RPS14 | |
The immunogen this antibody was made to, maps to a region between residue 101 to 151 of human Ribosomal Protein S14 using the numbering given in entry NP_001020242.1 (GeneID 6208). | |
100 μL | |
metabolism | |
6208 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction