Learn More
Invitrogen™ RPS15 Polyclonal Antibody

Description
Antibody detects endogenous levels of total RPS15.
Specifications
Specifications
| Antigen | RPS15 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RPS15 |
| Gene Accession No. | P62841, P62843, P62845 |
| Gene Alias | 40S ribosomal protein S15; homolog of rat insulinoma; insulinoma; insulinoma protein; ribosomal protein S15; rig; RIG protein; RPS15; S15; Small ribosomal subunit protein uS19 |
| Gene Symbols | RPS15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.