Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540733
Description
Peptide sequence: SKGPLQSVQV FGRKKTATAV AHCKRGNGLI KVNGRPLEMI EPRTLQYKLL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S9P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS16 | |
| Polyclonal | |
| Unconjugated | |
| RPS16 | |
| 40S ribosomal protein S16; AA420385; AI317031; im:6906552; ribosomal protein S16; Rps16; S16; si:dkey-66d18.1; similar to 40S ribosomal protein S16; Small ribosomal subunit protein uS9; zgc:153404 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6217 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62249 | |
| RPS16 | |
| Synthetic peptide directed towards the N-terminal of human RPS16 (aa 3-52). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction