Learn More
Invitrogen™ RPS16 Polyclonal Antibody

Description
Immunogen sequence: MPSKGPLQSV QVFGRKKTAT AVAHCKRGNG LIKVNGRPLE MIEPRTLQYK LLEPVLLLGK ERFAG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | RPS16 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RPS16 |
| Gene Accession No. | P62249 |
| Gene Alias | 40S ribosomal protein S16; AA420385; AI317031; im:6906552; ribosomal protein S16; Rps16; S16; si:dkey-66d18.1; similar to 40S ribosomal protein S16; Small ribosomal subunit protein uS9; zgc:153404 |
| Gene Symbols | RPS16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.