Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS17 Polyclonal Antibody
RPS17 Polyclonal Antibody
Supplier: Thermo Scientific PA575600
Description
RPS17 Polyclonal Antibody for Western Blot

Specifications
RPS17 | |
Polyclonal | |
Unconjugated | |
Rps17 | |
40S ribosomal protein S17; DBA4; ribosomal protein S17; Rps17; RPS17L; RPS17L1; RPS17L2; S17; Small ribosomal subunit protein eS17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20068, 29286, 6218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P04644, P08708, P63276 | |
Rps17 | |
Synthetic peptide corresponding to residues in Human RPS17. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction