Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563059
Description
Immunogen sequence: RGISIKLQEE ERERRDNYVP EVSALDQEII EVDPDTKEML KL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein S17, also known as RPS17, RPS17L1 or RPS17L2, is a 135 amino acid protein that is a component of the 40S subunit. Localized to the cytoplasm and expressed ubiquitously, Ribosomal Protein S17 belongs to the S17e family of ribosomal proteins and functions in protein synthesis. Mutations in the gene encoding Ribosomal Protein S17 are associated with Diamond-Blackfan anemia (DBA), a rare congenital disorder characterized by defective differentiation of pro-erythroblasts. Like most ribosomal proteins, Ribosomal Protein S17 exists as multiple processed pseudogenes that are scattered throughout the genome.
Specifications
RPS17 | |
Polyclonal | |
Unconjugated | |
Rps17 | |
40S ribosomal protein S17; DBA4; ribosomal protein S17; Rps17; RPS17L; RPS17L1; RPS17L2; S17; Small ribosomal subunit protein eS17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P08708 | |
Rps17 | |
Recombinant protein corresponding to Human RPS17. Recombinant protein control fragment (Product #RP-100843). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction