Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS19 Antibody (7E6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | RPS19 |
---|---|
Clone | 7E6 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
RPS19 Monoclonal antibody specifically detects RPS19 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
RPS19 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DBADBA1,40S ribosomal protein S19, ribosomal protein S19 | |
A synthesized peptide derived from Human RPS19 [UniProt P39019] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7E6 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6223 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title