Learn More
Invitrogen™ RPS19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596036
Description
Immunogen sequence: MPGVTVKDVN QQEFVRALAA FLKKSGKLKV PEWVDTVKLA KHKELAPYDE NWFYTRAAST ARHLYLRGGA GVGSMTKIYG GRQRNGVMPS HFSRGSKSVA RRVLQALEGL KMVEKDQDGG RKLTPQGQRD LDRIAGQVAA ANKKH; Positive Samples: HL-60, PC-3, MCF7, A-549, 293T, HepG2, SW620, BxPC-3; Cellular Location: Nucleus.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19E family of ribosomal proteins. It is located in the cytoplasm. Mutations in this gene cause Diamond-Blackfan anemia, a constitutional erythroblastopenia characterized by absent or decreased erythroid precursors, in a subset of patients. This suggests a possible extra-ribosomal function for this gene in erythropoietic differentiation and proliferation, in addition to its ribosomal function. Higher expression levels of this gene in some primary colon carcinomas compared to matched normal colon tissues has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS19 | |
Polyclonal | |
Unconjugated | |
RPS19 | |
40S ribosomal protein S19; DBA; DBA1; Dsk3; HGNC:10402; ribosomal protein S19; Rps19; S19; Small ribosomal subunit protein eS19 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6223 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P39019 | |
RPS19 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human RPS19 (NP_0010131). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.