Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS19BP1/AROS Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501569655
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9655 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9655 Supplier Bethyl Laboratories, Inc Supplier No. A305144AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

AROS, a nucleolar protein with an unknown function, interacts with extraribosomal protein RPS19, playing a role in the signaling pathways that regulate rRNA transcription. The deduced amino acid sequence of AROS, derived from cDNA, defines an isoelectric point of 11.3. No known functional motifs were found in AROS except a short polylysine tract embedded in a putative nucleolar localization signal. At a basal level the protein is expressed ubiquitously with a significantly high level of expression in some tissues. Human AROS gene is mapped to chromosome 22q13.1.

Specifications

Antigen RPS19BP1/AROS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RPS19BP1
Gene Accession No. Q86WX3
Gene Alias RP5-1104E15.4, AROS, S19BP, dJ1104E15.4, 40S ribosomal protein S19-binding protein 1, RPS19-binding protein 1, active regulator of SIRT1, homolog of mouse S19 binding protein
Gene Symbols RPS19BP1
Host Species Rabbit
Immunogen Between 86 and 136
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91582
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.