Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214853C
Description
RPS2 Polyclonal antibody specifically detects RPS2 in Human, Mouse samples. It is validated for Western BlotSpecifications
RPS2 | |
Polyclonal | |
Western Blot | |
LLREP3, MGC102851, MGC117344, MGC117345,40S ribosomal protein S4, OK/KNS-cl.6,40S ribosomal protein S2, Protein LLRep3, ribosomal protein S2, RPS4 | |
This antibody maps to a region between residue 243 and 293 of human Ribosomal Protein S2 using the numbering given in entry NP_002943.2 (GeneID 6187). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
6187 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction