Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599706 Supplier Invitrogen™ Supplier No. PA599706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Ribosomal Protein S2.

Bacterial pathogens deliver type III effector proteins into the plant cell during infection. Plants express disease resistance (R) proteins that respond specifically to a particular type III effector by activating immune responses. Resistance proteins guard the plant against pathogens that contain an appropriate avirulence protein via an indirect interaction. RPS2 (resistance to Pseudomonas syringae protein 2) is a 909 amino acid plasma membrane protein with leucine-rich repeat, leucine zipper and P loop domains that confers resistance to Pseudomonas syringae infection by interacting with the avirulence gene AvrRpt2. Belonging to the disease resistance NB-LRR family, RPS2 directly associates with RIN4, which triggers plant resistance when RIN4 is degraded by AvrRpt2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rps2
Gene Accession No. P15880, P27952
Gene Alias 40S ribosomal protein S2; 40S ribosomal protein S4; fc94f01; hypothetical protein LOC406565; ik:tdsubc_2h2; Llrep3; OK/KNS-cl.6; protein LLRep3; repeat family 3; ribosomal protein S2; ribosomal protein S2e; rps2; rps2e; Rps2r1; RPS4; S2; S4; Small ribosomal subunit protein uS5; tdsubc_2h2; wu:fb10e01; wu:fc94f01; wu:fj09b04; xx:tdsubc_2h2; zgc:85824
Gene Symbols Rps2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPS2(Accession P15880), corresponding to amino acid residues K257-T293.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6187, 83789
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.