Learn More
Invitrogen™ RPS20 Recombinant Rabbit Monoclonal Antibody (9H1D4)

Description
RPS20 Recombinant Monoclonal Antibody for ELISA, ICC/IF, IHC (P), IP, Western Blot
Specifications
Specifications
| Antigen | RPS20 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 9H1D4 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPS20 |
| Gene Accession No. | P60866, P60867, P60868 |
| Gene Alias | 40S ribosomal protein S20; 4632426K06Rik; AFAR; AFAR1; AKR7; Dsk4; ribosomal protein S20; Rps20; S20; Small ribosomal subunit protein uS10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.