Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS20 Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297654AF405
Description
RPS20 Polyclonal specifically detects RPS20 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RPS20 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 405 | |
40S ribosomal protein S20, FLJ27451, MGC102930, ribosomal protein S20 | |
Produced in rabbits immunized with E. coli-derived Human RPS20 fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
6224 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction